Learn More
R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF, Animal-Free Protein
Description
Specifications
Specifications
| Accession Number | NP_001997 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl. |
| Gene ID (Entrez) | 2247 |
| Molecular Weight (g/mol) | M.W.-theoretical: 16 kDa |
| Quantity | 1 mg |
| Source | E. coli-derived human FGF basic/FGF2/bFGF protein Pro143-Ser288 Produced using non-animal reagents in an animal-free laboratory. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.01 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.