Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF GMP Protein, CF

Catalog No. RD10008961
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10008961 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10008961 Supplier R&D Systems™ Supplier No. BTFGFBGMP025
Add to Cart
Add to Cart

Recombinant Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.100-1.00 ng/mL. The specific activity of Recombinant Human FGF basic/FGF2 GMP is >8.0 x 105 IU/mg, which is calibrated against the human FGF basic/FGF2 WHO International Standard (NIBSC code: 90/712).

Specifications

Accession Number NP_001997.5
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HEPES and Sodium Sulfate with Trehalose.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) Observed: 18 kDa, under reducing conditions., Observed: 17 kDa
Quantity 25 μg
Source E. coli-derived human FGF basic/FGF2/bFGF protein Ala135 - Ser288 Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤ -20°C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Regulatory Status GMP
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute 25 μg size at 250 μg/mL and all other sizes at 500 μg/mL in sterile water.
Purity or Quality Grade >97%, by SDS-PAGE with quantitative densitometry by Coomassie™ Blue Staining. The primary molecular weight by mass spectrometry is 17112 Da ± 10 Da
Protein FGF basic/FGF2/bFGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.