Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGFR1 beta (IIIb) Fc Chimera Protein

Catalog No. 765FR050
Change view
Click to view available options
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
765FR050 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 765FR050 Supplier R&D Systems™ Supplier No. 765FR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human FGF R1 beta (IIIb) Fc Chimera Protein is derived from NS0. The Recombinant Human FGF R1 beta (IIIb) Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2260
Molecular Weight (g/mol) 56 kDa (monomer)
Name FGFR1 beta
Quantity 50μg
Source Mouse myeloma cell line,NS0-derived human FGF R1 beta protein, (N-terminus) Human FGF R1 beta (IIIb) (Met1-Lys221) Accession &Num; NP_075594 47 aa IIIb Ig domain (His1-Pro47) Accession &Num; AAB19502 Human FGF R1 beta (IIIb) (Ala270-Glu285) Accession &Num; NP_075594 IEGRDMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.