Learn More
Creative Biomart Sprifermin, Recombinant human fibroblast growth factor 18 (rhFGF18) Protein
Supplier:  Creative Biomart THP0305
Encompass_Preferred
Sprifermin (developmental code name AS-902330), also known as recombinant human fibroblast growth factor 18 (rhFGF18), is a recombinant form of human fibroblast growth factor 18 (FGF18) which is under development by Merck and Nordic Bioscience for the treatment of osteoarthritis. As a recombinant form of FGF18, sprifermin is a potent agonist of the FGFR2 and FGFR3.
Catalog No. 50-208-4355