Learn More
R&D Systems™ Recombinant Human Fibronectin, ACFP Protein
Description
- Recommended concentration for this effect is typically 1 to 5 μg/cm2.
- Fibronectin can also be added to the media to support cell spreading at a concentration of 0.5 to 50 μg/mL.
- Optimal concentrations will need to be determined for individual user applications
Specifications
Specifications
| Accession Number | CAE45932 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 2335 |
| Molecular Weight (g/mol) | 180 to 250 kDa, reducing conditions |
| Quantity | 1 mg |
| Source | Spodoptera frugiperda, Sf 9 (baculovirus)-derived human Fibronectin protein Met1-Pro1908, with a C-terminal 6-His tag Produced in an animal component free process (ACFP) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.