Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fibronectin/Anastellin Protein

Catalog No. 8258FN050
Change view
Click to view available options
50ug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
82-58F-N050 50ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 82-58F-N050 Supplier R&D Systems™ Supplier No. 8258FN050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Fibronectin/Anastellin Protein is derived from E. coli. The Recombinant Human Fibronectin/Anastellin Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature. Upon receipt, store it immediately

Specifications

Accession Number P02751
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Molecular Weight (g/mol) 9 kDa
Name Fibronectin/Anastellin, Carrier Free
Preparation Method Reconstitute at 250μg/mL in PBS.
Quantity 50μg
Source E. coli-derived human Fibronectin/Anastellin protein Asn631-Pro705,with an N-terminal Met and a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Form Powder
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.