Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fibulin 5 Protein

Catalog No. 9006FB050
Change view
Click to view available options
50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
90-06F-B050 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-06F-B050 Supplier R&D Systems™ Supplier No. 9006FB050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Fibulin 5 Protein is derived from HEK293. The Recombinant Human Fibulin 5 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9UBX5
For Use With (Application) Bioactivity
Gene ID (Entrez) 10516
Molecular Weight (g/mol) Observed M.W.: 60-72kDa, reducing conditions, Theoretical M.W.: 49kDa
Quantity 50μg
Source Human embryonic kidney cell,HEK293-derived human Fibulin 5/DANCE protein Gln24-Phe448,with an N-terminal HA tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias ARMD3fibulin-5, Dance, DANCEFIBL-5, Developmental arteries and neural crest EGF-like protein, EVEC, FBLN5, fibulin 5, UP50, UP50FLJ90059, Urine p50 protein
Common Name Fibulin 5/DANCE
Gene Symbol FBLN5
Biological Activity Measured by the ability of the immobilized protein to enhance the adhesion of HUVEC human umbilical vein endothelial cells. The ED50 for this effect is typically 15-90ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Gln24-Phe448, with an N-terminal HA tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.