Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L Protein

Catalog No. p-4719680
Change view
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
308FK005CF 5 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
308FK025 25 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
308FK025CF 25 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 308FK005CF Supplier R&D Systems™ Supplier No. 308FK005/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is 0.2-1 ng/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 2323
Molecular Weight (g/mol) 17.5 kDa
Name Flt-3 Ligand
Quantity 5 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Flt-3 Ligand/FLT3L protein Thr27-Pro185
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.