Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human FLT3 protein, HEK293 hFc fusion | 100 UG
SDP

Catalog No. 5000292891
Encompass_Preferred

Recombinant human FLT3 extracellular domain produced in HEK293 cells and fused to a human Fc tag. Supplied lyophilized, the protein is intended for ligand-binding assays, ELISA, and receptor-ligand interaction studies; it includes storage and reconstitution recommendations and low endotoxin for sensitive assays.

  • Expression system: HEK293 cells.
  • Tag: human Fc fusion.
  • Formulation: lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Endotoxin level: <1 EU/μg.
  • Apparent molecular weight: 100-145 kDa.
  • Storage: store at -20°C; aliquot and freeze for long-term storage.
  • Reconstitution guidance: do not reconstitute below 100 μg/mL.
  • Biological activity: ED50 = 15.44 ng/mL in functional ELISA.

Catalog No. 50-002-92891 Supplier Medchemexpress LLC Supplier No. HYP75184100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human FLT3 extracellular domain produced in HEK293 cells and fused to a human Fc tag. Supplied lyophilized, the protein is intended for ligand-binding assays, ELISA, and receptor-ligand interaction studies; it includes storage and reconstitution recommendations and low endotoxin for sensitive assays.

  • Expression system: HEK293 cells.
  • Tag: human Fc fusion.
  • Formulation: lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Endotoxin level: <1 EU/μg.
  • Apparent molecular weight: 100-145 kDa.
  • Storage: store at -20°C; aliquot and freeze for long-term storage.
  • Reconstitution guidance: do not reconstitute below 100 μg/mL.
  • Biological activity: ED50 = 15.44 ng/mL in functional ELISA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.