Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fucosyltransferase 3/FUT3 Protein

Catalog No. 4950GT020
Change view
Click to view available options
20ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
4950GT020 20ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4950GT020 Supplier R&D Systems™ Supplier No. 4950GT020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

FUT3 has been formulated so that it can be used in the cell surface glycoengineering of living cells, and does not affect cell viability or native phenotype apart from the intended impact on cell glycobiology.

Measured by its ability to transfer fucose from GDP-fucose to N -Acetyllactosamine The specific activity is >25 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P21217
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 2525
Molecular Weight (g/mol) M.W. (Observed): 41 kDa, reducing conditions; M.W. (theoretical): 39 kDa
Name Fucosyltransferase 3/FUT3
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human Fucosyltransferase 3/FUT3 protein Arg35-Thr361, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol FUT3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Fucosyltransferase 3/FUT3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.