Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Galectin-9 Protein

Catalog No. 2045GA050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2045GA050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2045GA050 Supplier R&D Systems™ Supplier No. 2045GA050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

HEK293 rh Galectin-9 (Catalog # 9064-GA) is also available. TIM-3 binding is weaker than the E. coli version!

Measured by its ability to induce apoptosis of Jurkat human acute T cell leukemia cells. Lu, L.H. et al. (2007) J. Biochem. 141:157. The ED50 for this effect is 1-5 μg/mL. Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human Galectin-9 at 500 ng/mL can bind Recombinant Human TIM-3 Fc Chimera (Catalog # 2365-TM ) with an apparent Kd <30 nM.

Specifications

Accession Number BAA31542
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl, EDTA, DTT and Trehalose.
Gene ID (Entrez) 3965
Molecular Weight (g/mol) M.W. (Observed): 34 kDa, reducing conditions; M.W. (theoretical): 35.8 kDa
Name Galectin-9
Quantity 50 μg
Source E. coli-derived human Galectin-9 protein Ala2-Thr323
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Structural Form Monomer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL aq. soln..
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Galectin-9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.