Learn More
Biomatik Corporation Recombinant Human Gasdermin-D (GSDMD)
Supplier:  Biomatik Corporation RPC27181
Encompass_Preferred
Recombinant Human Gasdermin-D (GSDMD) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GSDMD. Target Synonyms: Gasdermin-D(Gasdermin domain-containing protein 1) [Cleaved into: Gasdermin-D, N-terminal(GSDMD-NT; hGSDMD-NTD); Gasdermin-D, C-terminal(GSDMD-CT; hGSDMD-CTD)]. Accession Number: P57764. Expression Region: 1~484aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 56.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7924