Learn More
Biomatik Corporation Recombinant Human Gastric inhibitory polypeptide receptor (GIPR), partial
Supplier:  Biomatik Corporation RPC27114
Encompass_Preferred
Recombinant Human Gastric inhibitory polypeptide receptor (GIPR), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GIPR. Target Synonyms: Glucose-dependent insulinotropic polypeptide receptor(). Accession Number: P48546. Expression Region: 22~138aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 18.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7925