Learn More
Creative Biomart GMP Grade Recombinant Human GBA protein
Supplier:  Creative Biomart GBA024THP
Encompass_Preferred
The protein is a hydrolytic lysosomal glucocerebroside-specific enzyme, which is a recombinant form of glucocerebrosidase indicated as a long-term enzyme replacement therapy for those suffering of Gaucher disease Type 1. It has an identical amino acid sequence to the naturally occurring enzyme. It is used for the treatment of Type 1 Gaucher disease, caused by a deficiency of the lysosomal enzyme glucocerebrosidase. Additionally, Velaglucerase alfa has also been investigated for use in Type 3 Gaucher disease.
Catalog No. 50-208-5357