Learn More
Abcam Recombinant Human GDF15 protein, 100UG
Supplier:  Abcam AB125769100UG
Encompass_Preferred
Recombinant Human GDF15 protein is a Human Fragment protein, in the 197 to 308 aa range, expressed in Escherichia coli, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: MIC1, PDF, PLAB, PTGFB, GDF15, Growth/differentiation factor 15, GDF-15, Macrophage inhibitory cytokine 1, NSAID-activated gene 1 protein, NSAID-regulated gene 1 protein, Placental TGF-beta, Placental bone morphogenetic protein, Prostate differentiation factor, MIC-1, NAG-1, NRG-1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4852