Learn More
R&D Systems™ Recombinant Human GITR/TNFRSF18 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q9Y5U5 |
| For Use With (Application) | Binding Activity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 8784 |
| Molecular Weight (g/mol) | M.W. (Observed): 50 kDa, reducing conditions; M.W. (theoretical): 41 kDa (monomer) |
| Name | GITR/TNFRSF18 Fc Chimera |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line, NS0-derived human GITR/TNFRSF18 protein Human GITR (Gln26-Glu161) (Thr45Ala) (N-terminus) Accession # Q9Y5U5 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.