Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human glucokinase (GCK) protein | 9001-51-8 | >95.0% | 5 UG
SDP

Catalog No. 5000294576
Encompass_Preferred

Recombinant human glucokinase (GCK) protein supplied as a 5 μg research-grade enzyme. GCK catalyzes phosphorylation of D-glucose to glucose-6-phosphate and acts as a glucose sensor in pancreatic beta cells and the liver; the product is intended for in vitro research applications and should be handled and stored under recommended conditions.

  • Recombinant human enzyme corresponding to amino acids L2-Q465.
  • Expressed in Escherichia coli for recombinant production.
  • Tag free to reduce assay interference.
  • Purity >95% by reducing SDS-PAGE.
  • Formulated in Tris-HCl, NaCl, 10% glycerol; 0.22 μm filtered.
  • Store at -80°C and avoid repeated freeze-thaw cycles.
  • Endotoxin <1 EU/μg (LAL method).
  • Specific activity defined per unit at pH 8.0, 37°C.

Catalog No. 50-002-94576 Supplier Medchemexpress LLC Supplier No. HYP757905UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human glucokinase (GCK) protein supplied as a 5 μg research-grade enzyme. GCK catalyzes phosphorylation of D-glucose to glucose-6-phosphate and acts as a glucose sensor in pancreatic beta cells and the liver; the product is intended for in vitro research applications and should be handled and stored under recommended conditions.

  • Recombinant human enzyme corresponding to amino acids L2-Q465.
  • Expressed in Escherichia coli for recombinant production.
  • Tag free to reduce assay interference.
  • Purity >95% by reducing SDS-PAGE.
  • Formulated in Tris-HCl, NaCl, 10% glycerol; 0.22 μm filtered.
  • Store at -80°C and avoid repeated freeze-thaw cycles.
  • Endotoxin <1 EU/μg (LAL method).
  • Specific activity defined per unit at pH 8.0, 37°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.