Learn More
R&D Systems™ Recombinant Human Glypican 3 His Alexa Fluor™ 647 Protein
Description
Specifications
Specifications
| Accession Number | P51654.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 2719 |
| Molecular Weight (g/mol) | M.W.-observed: 60-100 kDa, under non-reducing conditions., M.W.-theroretical: 62 kDa |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human Glypican 3 protein Gln25-His559 with a C-terminal 6-His tag |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GPC3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.