Learn More
Proteintech Group Inc Recombinant Human GM-CSF 100ug
SDP
Supplier:  Proteintech Group Inc HZ1002100UG
Encompass_Preferred
Animal-free Recombinant Human GM-CSF is produced in a human cell expression system in serum-free, chemically defined media. It has a demonstrated greater stability under cell culture conditions making it ideal for efficient generation of Human Dendritic Cells. GM-CSF is a hematopoietic growth factor that stimulates the development of early erythroid megakaryocytic and eosinophilic progenitor cells. It is produced in endothelial cells, monocytes, fibroblasts, and T lymphocytes. Recombinant Human GM-CSF is a 15 to 36 kDa golbular protein consisting of 128 amino acids, containing two intramolecular disulfide bonds and two potential N-linked glycosylation sites.
Catalog No. 50-197-3837