Learn More
Creative Biomart Recombinant Human GPC3 protein, His-tag, GMP Grade
Supplier:  Creative Biomart GPC3551THP
Encompass_Preferred
Recombinant Human GPC3 protein GMP Grade(NP_004475.1)(534 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5300