Learn More
Abcam Recombinant human GRP78 BiP protein (Active), 50UG
Supplier:  Abcam AB7843250UG
Encompass_Preferred
Recombinant human GRP78 BiP protein (Active) is a Human Full Length protein, in the 1 to 654 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, ELISA, CBA, WB, FuncS.
Protein/ Peptide/ Enzyme Synonyms: GRP78, HSPA5, Endoplasmic reticulum chaperone BiP, 78 kDa glucose-regulated protein, Binding-immunoglobulin protein, Heat shock protein 70 family protein 5, Heat shock protein family A member 5, Immunoglobulin heavy chain-binding protein, GRP-78, BiP, HSP70 family protein 5
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1200779