Learn More
Medchemexpress LLC Recombinant human guanylate-binding protein 1 (GBP1) | >95.0% | 10 UG
SDP
Catalog No. 5000293078
Encompass_Preferred
Recombinant human guanylate-binding protein 1 (GBP1) expressed in HEK293 cells with a C-terminal His tag. GBP1 is a dynamin-family large GTPase involved in innate immunity; it hydrolyzes GTP to GMP and contributes to inflammasome regulation. The product is supplied as a filtered PBS solution, is highly pure, and is provided in microgram quantities for biochemical and cell-based research.
Catalog No. 50-002-93078 Supplier Medchemexpress LLC Supplier No. HYP7517210UG
Recombinant human guanylate-binding protein 1 (GBP1) expressed in HEK293 cells with a C-terminal His tag. GBP1 is a dynamin-family large GTPase involved in innate immunity; it hydrolyzes GTP to GMP and contributes to inflammasome regulation. The product is supplied as a filtered PBS solution, is highly pure, and is provided in microgram quantities for biochemical and cell-based research.