Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HGF (NS0 Expressed) Protein

Catalog No. 11352HG010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-352-HG010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-352-HG010 Supplier R&D Systems™ Supplier No. 11352HG010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wild type

Measured by its ability to induce IL-11 secretion by Saos-2 human osteosarcoma cells. Hjertner, O. et al. (1999) Blood 94:3883. The ED50 for this effect is equal to or less than 4.00 ng/mL.

Specifications

Accession Number P14210.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose.
Gene ID (Entrez) 3082
Molecular Weight (g/mol) M.W. (Observed): 30-40 kDa ( beta chain) & 54-66 kDa ( alpha chain), under reducing conditions., M.W. (Theoretical): 54 kDa ( alpha chain) & 26 kDa ( beta chain)
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human HGF protein Gln32-Arg494 alpha chain & Val495-Ser728 beta chain
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.50 EU per 1μg of the protein by the LAL method.
Gene Alias deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute 10μg size at 100μg/mL in PBS. Reconstitute all the other sizes at 200μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein HGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.