Learn More
Abcam Recombinant Human HIF-1 alpha protein, 50UG
Supplier:  Abcam AB15447850UG
Encompass_Preferred
Recombinant Human HIF-1 alpha protein is a Human Full Length protein, in the 1 to 735 aa range, expressed in Escherichia coli, with >75% purity and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: BHLHE78, MOP1, PASD8, HIF1A, Hypoxia-inducible factor 1-alpha, HIF-1-alpha, HIF1-alpha, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, Class E basic helix-loop-helix protein 78, Member of PAS protein 1, PAS domain-containing protein 8, bHLHe78
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1488240