Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-alpha-Synuclein Protein

Catalog No. SP480500
Change view
Click to view available options
500ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SP480500 500ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SP480500 Supplier R&D Systems™ Supplier No. SP480500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human His6-Synuclein-alpha Protein is derived from E. coli. The Recombinant Human His6-Synuclein-alpha Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P37840.1
Formulation Supplied as a 0.2 μm filtered solution in HEPES and NaCl.
Gene ID (Entrez) 6622
Molecular Weight (g/mol) 15 kDa
Name His6 Synuclein-alpha
Quantity 500μg
Source E. coli-derived human alpha-Synuclein protein Met1 - Ala140 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.