Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His8-(DYKDDDDK)-BRD4 (49-460) Protein

Catalog No. SP600100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SP600100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SP600100 Supplier R&D Systems™ Supplier No. SP600100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Accession Number O60885
For Use With (Application) Bioactivity
Gene ID (Entrez) 23476
Molecular Weight (g/mol) 52 kDa
Name bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1
Quantity 100 μg
Source E. coli-derived human BRD4 protein Glu49 - Glu460
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 60 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
Conjugate Unconjugated
Species Human
Buffer X mg/ml (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.