Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HSP70/HSPA1A Protein

Catalog No. AP100100
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP100100 100 μg
AP100050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AP100100 Supplier R&D Systems™ Supplier No. AP100100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Reaction conditions will need to be optimized for each specific application. We recommend an initial HSP70/HSPA1A concentration of 2-3 μM for in vitro use. IMPORTANT: HSP40/DNAJB1 (Catalog # AP-110), or another suitable co-chaperone, is required for HSP70/HSPA1A activity and should be used at a concentration that is equimolar to HSP70/HSPA1A.

Specifications

Formulation X mg/mL (X μM) in 50 mM HEPES pH 8, 100 mM NaCl, 5 mM DTT
Gene ID (Entrez) 3303
Molecular Weight (g/mol) 70 kDa
Name HSP70/HSPA1A
Quantity 100 μg
Source E. coli-derived human HSP70/HSPA1A protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Structural Form Solution
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.