Learn More
R&D Systems™ Recombinant Human HVEM/TNFRSF14 Fc Alexa Fluor™ 488 Protein
Description
Specifications
Specifications
| Accession Number | Q92956.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 8764 |
| Molecular Weight (g/mol) | M.W.-observed: 55-75 kDa, under reducing conditions., M.W.-theroretical: 44 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human HVEM/TNFRSF14 protein Human HVEM/TNFRSF14 (Pro37-Val202) (C-terminus) Accession # Q92956.3 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.