Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human IFN-γ

Catalog No. BDB554617 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
Concentration:
250μg/mL
500μg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
BDB554617 50 μg 500μg/mL
BDB554616 25 μg 250μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB554617 Supplier BD Supplier No. 554617
Add to Cart
Add to Cart

Pure by SDS-PAGE, and an absorbance assay based on the Beers-Lambert law

Frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution and containing 2.5mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no azide

  • Reactivity: Human
  • Storage Buffer: Frozen aqueous buffered solution containing BSA
  • Biological Activity: Activity range is measured in an anti-viral bioassay using A549 indicator cells
  • Specific Activity: 0.1 - 2.0 x 108 Units/mg
  • ED50: 0.05 to 1.0 ng/ml
  • Observed dose response relationship: 16X

Bioassay, enzyme-linked immunosorbent assay standard

TRUSTED_SUSTAINABILITY

Specifications

Concentration 500μg/mL
For Use With (Application) Bioassay, ELISA Standard
Formulation Frozen aqueous Buffered solution containing BSA
Molecular Weight (g/mol) 14 to 18
Name Human IFN-gamma Recombinant Protein
Purification Method SDS-PAGE
Quantity 50 μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Solution
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.