Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IFN-alpha B2/IFNA8 Protein

Catalog No. 11018IF010
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11018IF010 10 μg
11018IF050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11018IF010 Supplier R&D Systems™ Supplier No. 11018IF010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is 1.20-12.0 pg/mL.

Specifications

Accession Number P32881.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3445
Molecular Weight (g/mol) MolecularWeight-observed: 23-27 kDa, under reducing conditions.; MolecularWeight-theroretical: 19 kDa
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived IFN-alpha B2/IFNA8 protein Cys24-Glu189
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IFNA8, IFNalpha B2, IFN-alpha B2, IFN-alpha-8, IFN-alphaB, interferon alpha type 201, interferon alpha-8, interferon alpha-B', Interferon alpha-B, Interferon alpha-B2, interferon, alpha 8, LeIF B
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IFN-alpha B2/IFN-alpha 8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.