Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IFN-alpha/beta R1 Fc Chimera Protein, CF

Catalog No. 11574AB050
Change view
Click to view available options
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-574-AB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-574-AB050 Supplier R&D Systems™ Supplier No. 11574AB050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Measured by its binding ability in a functional ELISA. Recombinant Human IFN-alpha/beta R1 Fc Chimera (Catalog # 11574-AB) binds Recombinant Human LRPAP (Catalog # 4296-LR ) with an ED50 of 0.750-9.00 μg/mL.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3454
Molecular Weight (g/mol) MolecularWeight-observed: 105-120 kDa, under reducing conditions., MolecularWeight-theroretical: 74 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human IFN-alpha/beta R1 protein Human IFNAR1 (Lys28-Lys436) (N-terminus) Accession # AAA52730.1 GGIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFN-aR1, IFNAR1, IFNBR, IFNbR1, IFN-bR1, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water.
Protein IFN-alpha/beta R1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.