Learn More
Apexbio Technology LLC Recombinant Human IFN-beta 100ug
SDP
Catalog No. 5000094930
Encompass_Preferred
Recombinant Human IFN-beta is a recombinant immunomodulatory protein targeting the type I interferon signaling pathway It is designed to bind the IFN- / receptor complex thereby activating the JAK/STAT signaling cascade and modulating inflammatory responses including regulation of Th17 cell activity Recombinant Human IFN-beta exerts its biological activity primarily through activation of JAK/STAT signaling upon receptor engagement In animal models deficiency of IFN-beta leads to increased susceptibility to experimental autoimmune encephalomyelitis (EAE) a model for human multiple sclerosis Based on these pharmacological properties Recombinant Human IFN-beta holds research potential in investigating autoimmune and inflammatory diseases and antiviral therapies
Catalog No. 50-000-94930 Supplier Apexbio Technology LLC Supplier No. PH20430.1
Recombinant Human IFN-beta is a recombinant immunomodulatory protein targeting the type I interferon signaling pathway It is designed to bind the IFN- / receptor complex thereby activating the JAK/STAT signaling cascade and modulating inflammatory responses including regulation of Th17 cell activity Recombinant Human IFN-beta exerts its biological activity primarily through activation of JAK/STAT signaling upon receptor engagement In animal models deficiency of IFN-beta leads to increased susceptibility to experimental autoimmune encephalomyelitis (EAE) a model for human multiple sclerosis Based on these pharmacological properties Recombinant Human IFN-beta holds research potential in investigating autoimmune and inflammatory diseases and antiviral therapies