Learn More
R&D Systems™ Recombinant Human IFN-omega Protein
Description
Specifications
Specifications
| Accession Number | X58822 |
| For Use With (Application) | Bioactivity |
| Formulation | Phosphate buffered saline (PBS) containing 2% bovine serum albumin (BSA) |
| Molecular Weight (g/mol) | Predicted MW: 20kDa |
| Source | E. coli-derived human IFN-omega protein |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.