Learn More
R&D Systems™ Recombinant Human IGF-I R/IGF1R Fc Chimera Protein, CF
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3480 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 160-180 kDa, 110-125 kDa and 70-80 kDa, under reducing conditions, MolecularWeight-theroretical: 130 kDa (single chain), 80 kDa (alpha subunit) and 49 kDa (beta subunit) |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human IGF-I R/IGF1R protein Human IGF1R (Glu31-His935) (N-terminus) Accession # P08069.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.