Learn More
AbD Serotec Recombinant Human IgG2 Lambda, 0.1mg
Supplier:  AbD Serotec HCA108
Encompass
Recombinant Human IgG2 lambda is a recombinant antibody with specificity for Green Fluorescent Protein. It has no known reactivity with mammalian proteins or other antigens. It is therefore recommended as a control reagent for assays where this immunoglobulin is measured, or when using other human antibodies of the same isotype and subclass. It is recommended that this reagent is used at the same concentration as the test reagent.
Catalog No. NC1616533