Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-10 Protein

Catalog No. 217ILB100CF
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-7IL-B100CF 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-7IL-B100CF Supplier R&D Systems™ Supplier No. 217ILB100/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured in a cell proliferation assay using MC/9-2 mouse mast cells. The ED50 for this effect is 0.15 to 0.75 ng/mL.

Specifications

Accession Number P22301
For Use With (Application) Bioactivity
Gene ID (Entrez) 3586
Molecular Weight (g/mol) 18.6 kDa
Name CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Quantity 100 μg
Source Spodoptera frugiperda, Sf 21 (stably transfected)-derived human IL-10 protein Met1-Asn178
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol IL10
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute 5 μg vials at 50 μg/mL in sterile PBS. Reconstitute 25 μg or larger vials at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <1.0 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.