Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-10 R alpha Fc Chimera Protein

Catalog No. 9044RI050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-44R-I050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-44R-I050 Supplier R&D Systems™ Supplier No. 9044RI050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-10 R alpha Fc Chimera Protein is derived from HEK293. The Recombinant Human IL-10 R alpha Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q13651
For Use With (Application) Bioactivity
Gene ID (Entrez) 3587
Molecular Weight (g/mol) Observed M.W.: 64-78kDa, reducing conditions, Theoretical M.W.: 51kDa
Name IL-10R alpha
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human IL-10 R alpha protein, (N-terminus) Human IL-10 R alpha (His22-Asn235) Accession &Num; Q13651 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL10R alpha, IL-10R subunit 1, IL-10R1, IL10RA, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Common Name IL-10 R alpha
Gene Symbol IL10RA
Biological Activity Measured by its ability to inhibit IL-10-dependent proliferation of MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. The ED50 for this effect is typically 6-36ng/mL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human IL-10 R alpha (His22-Asn235) Accession # Q13651, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.