Learn More
Proteintech Group Inc Recombinant Human IL-17F 10ug
SDP
Supplier:  Proteintech Group Inc HZ111610UG
Encompass_Preferred
Animal-free Recombinant Human IL-17F is a homodimeric glycoprotein with an apparent molecular mass of 38 kDa. This cytokine is produced in a human cell expression system with serum-free, chemically defined media. Production in human 293 cells offers authentic glycosylation, which contributes to stability in cell growth media and other applications. IL-17 is commonly associated with allergic responses. IL-17 induces the production of many other cytokines (such as IL-6, G-CSF, GM-CSF, IL-1beta, TGF beta, and TNF alpha), chemokines (including IL-8, GRO alpha, and MCP-1), and prostaglandins (e.g., PGE2) from many cell types (fibroblasts, endothelial cell, epithelial cells, keratinocytes, and macrophages). The release of cytokines causes many functions, such as airway remodeling, a characteristic of IL-17 responses.
Catalog No. 50-197-3894