Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-18 R beta/IL-1 R7 Fc Avi Protein

Catalog No. AVI118050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AVI118050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AVI118050 Supplier R&D Systems™ Supplier No. AVI118050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Biotinylated

Measured by its binding ability in a functional ELISA. When Recombinant Human IL-18 R alpha/IL-1 R5 Fc Chimera (Catalog # 816-LR ) is coated at 1.00 μg/mL (100 μL/well), in the presence of 50.0 ng/mL of Recombinant Human IL-18 (Catalog # 9124-IL ), Biotinylated Recombinant Human IL-18 R beta/IL-1 R7 Fc Chimera Avi-tag binds with an ED50 of 10.0-100 ng/mL. The biotin to protein ratio is greater than 0.7 as determined by the HABA assay.

Specifications

Accession Number O95256.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 8807
Molecular Weight (g/mol) M.W.-observed: 87-97 kDa, under reducing conditions., M.W.-theroretical: 65 kDa (monomer)
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL-18 R beta/IL-1 R7 protein Human IL-18 R beta/IL-1 R7(Phe20-Arg356)(N-terminus) Accession # O95256.1 DIEGRMD Human IgG1(Pro100-Lys330) Avi-tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Accessory protein-like, AcPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-1 R7, IL-18 receptor accessory protein, IL-18 receptor beta, IL18R beta, IL-18 RAcP, IL18RAP, IL-18 Rb, IL18RB, IL-18 Rbeta, IL-18 R-beta, IL-1 R accessory protein-like, IL1R7, IL-1R7, IL-1 R-7, IL-1 RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590
Conjugate Biotin
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-18 R beta/IL-1 R7/ACPL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.