Learn More
Proteintech Group Inc Recombinant Human IL-2 1000ug
SDP
Supplier:  Proteintech Group Inc HZ10151000UG
Encompass_Preferred
Animal-free Recombinant Human IL-2 is an O-glycosylated, four alpha -helix bundle cytokine. It is expressed by gamma T cells, CD4+ and CD8+ T cells, eosinophils, B cells, and dendritic cells. IL-2 has potent stimulatory activity for antigen-activated T cells. It is a monomeric glycoprotein with an apparent molecular mass of 15 kDa. This cytokine is produced in a human cell expression system with serum-free, chemically defined media. Production in human 293 cells offers authentic glycosylation, which contributes to stability in cell growth media.
Catalog No. 50-197-3847