Learn More
Eurofins Discoverx Recombinant Human IL-2 | Cell Culture Grade | 10ug
Supplier:  Eurofins Discoverx 921331
Encompass
Interleukin 2 (IL-2) is a cytokine protein and intricate member of the immune response. Regulates activation of T and B lymphocyte proliferation, and also stimulates monocytes, lymphokine-activated killer cells, natural killer cells, and glioma cells. This cell culture grade recombinant human IL-2 is a variant form of IL-2 (contains a serine to cysteine substitution) that can be used for differentiation studies, functional assays, and cell culture.
Catalog No. NC1413276