Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-2 (CD122-Directed) Protein, CF

Catalog No. BT002DBR500
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT002DBR500 500 μg
BT002DBR100 100 μg
BT002DBR050 50 μg
BT002DBR010 10 μg
BT002DBR01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. BT002DBR500 Supplier R&D Systems™ Supplier No. BT002DBR500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

AI Modified Protein

Measured in a cell proliferation assay using NK-92 human natural killer lymphoma cells. The ED50 for this effect is 0.0500-0.500 ng/mL.

Specifications

Accession Number P60568.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Sodium Acetate with Trehalose.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) M.W.-observed: 12-14 kDa, under reducing conditions., M.W.-theroretical: 15 kDa
Quantity 500 μg
Source E. coli-derived human IL-2 protein Ala21-Thr153 (C145S) with and without an N-terminal Met Proprietary point mutations
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.20 EU per 1 μg of the protein by the LAL method.
Gene Alias aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in water. Reconstitute all the other sizes at 500 μg/mL in water.
Protein IL-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.