Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-2 Heat Stable Agonist Protein, CF

Catalog No. BT002HS01M
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT002HS01M 1 mg
BT002HS100 100 μg
BT002HS010 10 μg
BT002HS050 50 μg
BT002HS500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. BT002HS01M Supplier R&D Systems™ Supplier No. BT002HS01M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

AI Modified Protein

Measured in a cell proliferation assay using NK-92 human natural killer lymphoma cells. The ED50 for this effect is 0.0150-0.150 ng/mL.

Specifications

Accession Number Engineered based on P60568.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) MolecularWeight-observed: 7 kDa, under reducing conditions., Protein: IL-2
Quantity 1 mg
Source E. coli-derived human IL-2 protein Proprietary
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Protein IL-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.