Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-2 (Mammalian-expressed) Protein

Catalog No. 10453IL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10453IL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10453IL050 Supplier R&D Systems™ Supplier No. 10453IL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. Gearing, A.J.H. and C.B. Bird (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 295.The ED50 for this effect is 0.05-0.3 ng/mL.

Specifications

Accession Number P60568.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) M.W. Observed: 14-18 kDa, under reducing conditions, M.W. Theoretical: 15 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL-2 protein Ala21-Thr153
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias aldesleukin, IL2, IL-2 lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Conjugate Unconjugated
Reconstitution Reconstitute at 100-500 μg/mL in sterile 100 mM Acetic Acid
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.