Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-2 Protein, Animal-Free

Catalog No. p-7250148
Change view
Click to view available options
Quantity:
1 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT002AFL01M 1 mg
BT002AFL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BT002AFL01M Supplier R&D Systems™ Supplier No. BT002AFL01M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains the Cysteine to Serine substitution found in Proleukin™ (aldesleukin)

Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. Gearing, A.J.H. and C.B. Bird (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 295.

The ED50 for this effect is 0.0300-0.250 ng/mL.

The specific activity of recombinant human IL-2 is >5.00 x 106 IU/mg, which is calibrated against the human IL-2 WHO International Standard (NIBSC code: 86/500).

Specifications

Accession Number P60568.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Sodium Acetate with Trehalose.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) M.W.-observed: 13 kDa, under reducing conditions., M.W.-theroretical: 15.5 kDa
Quantity 1 mg
Source E. coli-derived human IL-2 protein Ala21-Thr153 (Cys145Ser), with and without an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias aldesleukin, IL2, IL-2, IL-2 lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in sterile deionized water.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.