Learn More
Sino Biological Recombinant Human IL-20 Protein, Biotinylated 20
Catalog No. 5000698967
Encompass_Preferred
A DNA sequence encoding the mature form of human IL20 (Q9NYY1) (Leu25-Glu176) was expressed, with a N-terminal Met. The purified protein was biotinylated in vitro.
Catalog No. 50-006-98967 Supplier Sino Biological Supplier No. 13060HNAEB20
A DNA sequence encoding the mature form of human IL20 (Q9NYY1) (Leu25-Glu176) was expressed, with a N-terminal Met. The purified protein was biotinylated in vitro.