Learn More
R&D Systems™ Recombinant Human IL-21 Protein, Animal-Free
Description
Specifications
Specifications
| Accession Number | Q9HBE4.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 59067 |
| Molecular Weight (g/mol) | M.W.-observed: 15-17 kDa, under reducing conditions., M.W.-theroretical: 15 kDa |
| Quantity | 25 μg |
| Source | E. coli-derived human IL-21 protein Gln32-Ser162 with a N-terminal Met Produced using non-animal reagents in an animal-free laboratory. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.