Learn More
R&D Systems™ Recombinant Human IL-26/AK155 Dimer Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol. |
| Gene ID (Entrez) | 55801 |
| Molecular Weight (g/mol) | 17.7 kDa (monomer) |
| Name | IL-26/AK155 Dimer |
| Quantity | 10 μg |
| Source | E. coli-derived human IL-26/AK155 protein Lys22-Gln171,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Structural Form | Disulfide-linked homodimer and a small amount of monomer (no more than 10%) |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.