Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-28A/IFN-lambda 2 (HEK293) Protein

Catalog No. 8417IL025CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
84-17I-L025CF Lyophilized from a 0.2μm filtered solution in PBS
84-17I-L025 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 84-17I-L025CF Supplier R&D Systems™ Supplier No. 8417IL025/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-28A/IFN-lambda 2 (HEK293) Protein is derived from HEK293. The Recombinant Human IL-28A/IFN-lambda 2 (HEK293) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q8IZJ0
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 282616
Molecular Weight (g/mol) 20kDa
Name IL-28A/IFN-lambda 2
Preparation Method Reconstitute at 100μg/mL in PBS.
Quality Control Testing SDS-PAGE: 20-23kDa, reducing condtions
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human IL-28A/IFN-lambda 2 protein Val26-Val200
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Val26 & Val28
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.