Learn More
Proteintech Group Inc Recombinant Human IL-28B 10ug
SDP
Supplier:  Proteintech Group Inc HZ124510UG
Encompass_Preferred
Animal-free Recombinant Human IL-28B (IFN-lambda 3) is expressed in human 293 cells with an apparent molecular mass of 24 kDa. This cytokine is produced in a serum-free, chemically defined media. Production in human 293 cells offers authentic human post-translational modification which contributes to stability in cell growth media and other applications. The purity is greater than 95%.
Catalog No. 50-197-3941